missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VOC Alcohol Mixture 40000 μg/mL in Methanol, Dr. Ehrenstorfer
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
Specifications
Specifications
| CAS | 64-17-5, 67-63-0 |
| Quantity | 5 x 1 mL |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction