missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VO-OHpic
GREENER_CHOICE

Product Code. 18761399
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
missing translation for 'unitSize'
10mg
50mg
This item is not returnable. View return policy

Product Code. 18761399

missing translation for 'mfr': Tocris Bioscience™ 3591/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent PTEN inhibitor

VO-OHpic is a potent inhibitor of PTEN (phosphatase and tensin homolog) (IC50 = 35 nM). Displays selectivity for PTEN over cysteine-based phosphatases such as SopB, MTM, PTPβ and SAC (IC50 values are 588 nM, 4.03, 57.5 and >10 μM respectively). Increases PIP2 and PIP3 levels; also increases Akt translocation in NIH/3T3 fibroblasts. Blocks the immunosuppressant activity of IDO-activated Tregs. Promotes antitumor inflammatory response in combination with immunotherapy and enhances anticancer chemotherapy in tumor-bearing mice.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mg
Product Type VO-OHpic
Purity >98%
Molecular Weight (g/mol) 361.16
Inhibitors VO-OHpic

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.