missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VNN3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534459
This item is not returnable.
View return policy
Description
A suggested positive control is human brain tissue lysate. PA5-34459 can be used with blocking peptide PEP-1501.
The vanin family is a novel group of ectoenzymes that function in tissue repair and plays a role in oxidative-stress response. As both secreted and membrane proteins, the vanin family members have been implicated as therapeutic targets in inflammatory disease. VNN3 (vascular non-inflammatory molecule 3), also known as Vanin3, is a 501 amino acid GPI-anchored amidohydrolase that is widely expressed and is found at highest levels in blood and liver. Induced by Th17 / Th1 type cytokines, VNN3 converts pantetheine into pantothenic acid. Containing one CN hydrolase domain, VNN3 is encoded by a gene that maps to human chromosome 6q23.2.
Specifications
| VNN3 | |
| Polyclonal | |
| Unconjugated | |
| VNN3 | |
| HSA238982; pantetheinase pseudogene; pantetheinase-associated gene expressed in leukocytes (PAGEL)-alpha; RGD1560609; vanin 1 pseudogene; vanin 3; vanin-3; Vascular non-inflammatory molecule 3; VNN3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26464, 498992, 55350 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q8WZ42, Q9QZ25 | |
| VNN3 | |
| A 19 amino acid synthetic peptide near the carboxy terminus of human VNN3. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur