missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VNN2 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99866G
This item is not returnable.
View return policy
Description
VNN2 Polyclonal specifically detects VNN2 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| VNN2 | |
| Polyclonal | |
| Western Blot, ELISA | |
| EC 3.5.1, EC 3.5.1.92, FOAP-4, Glycosylphosphatidyl inositol-anchored protein GPI-80, GPI-80, GPI-80 variant protein 1, GPI-80 variant protein 2, GPI-80 variant protein 3, GPI-80 variant protein 4, pantetheinase, Protein FOAP-4, vanin 2, vanin-2, Vannin 2, vascular non-inflammatory molecule 2 | |
| Produced in rabbits immunized with purified, recombinant VNN2 (NP_004656.2; Met 1-Ser 492). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 8875 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction