missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VNN1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21745-1-AP-150UL
This item is not returnable.
View return policy
Description
Hematopoietic precursor cells migrate to the thymus, where they differentiate into mature T lymphocytes. GPI-anchored vanin-1 protein regulates the late adhesion steps of thymus homing of bone marrow precursor cells. Vanin-1 is ubiquitously expressed as a pantetheinase enzyme and catalyzes the hydrolysis of pantetheine for vitamin B5 recycling. The hydrolytic activity of vanin-1 generates the potent antioxidant cysteamine as a metabolite. As a membrane bound pantetheinase, vanin-1 provides the main source of cysteamine under normal physiological conditions. In mice, vanin-1 is expressed specifically in male Sertoli cells of the developing testis, where it aids in cell migration.Specifications
| VNN1 | |
| Polyclonal | |
| Unconjugated | |
| VNN1 | |
| HDLCQ8, Pantetheinase, Pantetheine hydrolase, Tiff66, vanin 1, VNN1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22361, 8876 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95497, Q9Z0K8 | |
| VNN1 | |
| VNN1 Fusion Protein Ag16501 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction