missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VN1R1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599936
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total VN1R1.
Pheromones are chemical signals that elicit specific behavioral responses and physiologic alterations in recipients of the same species. The protein encoded by this gene is similar to pheromone receptors and is primarily localized to the olfactory mucosa. An alternate splice variant of this gene is thought to exist, but its full length nature has not been determined. Putative pheromone receptor. Expressed in the olfactory mucosa, very low expression in brain, lung and kidney. The chimpanzee and orangutan orthologous proteins do not exist, their genes are pseudogenes.
Specifications
| VN1R1 | |
| Polyclonal | |
| Unconjugated | |
| VN1R1 | |
| G-protein coupled receptor GPCR24; hGPCR24; V1RL1; V1R-like 1; V1r-like receptor 1; V3r-related gene protein; VN1R1; VNR19I1; vomeronasal 1 receptor 1; vomeronasal olfactory receptor chromosome 19 subtype I member 1; vomeronasal olfactory receptor, (chromosome 19) subtype I, member 1; vomeronasal type-1 receptor 1; ZVNH1; ZVNR1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57191 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9GZP7 | |
| VN1R1 | |
| A synthesized peptide derived from human VN1R1(Accession Q9GZP7), corresponding to amino acid residues G3-L53. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction