missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VMO1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5148262
This item is not returnable.
View return policy
Description
VMO1 Polyclonal Antibody for IHC (P)
VMO1 (Vitelline Membrane Outer Layer 1 Homolog) is a Protein Coding gene.
Specifications
| VMO1 | |
| Polyclonal | |
| Unconjugated | |
| Vmo1 | |
| ERGA6350; ERGA6350 antibody; Gm741; MGC125880 antibody; MGC125881 antibody; PRO21055; PRO21055 antibody; RGD1559861; UNQ6350/PRO21055; vitelline membrane outer layer 1 homolog; vitelline membrane outer layer 1 homolog (chicken); vitelline membrane outer layer 1 homolog (chicken) antibody; vitelline membrane outer layer protein 1 homolog; vitelline membrane outer layer protein 1 homolog antibody; VMO1; VMO1 antibody | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 284013 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 100 μg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q7Z5L0 | |
| Vmo1 | |
| E. coli-derived Human VMO1 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction