missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VMAT2 Monoclonal Antibody (OTI10C11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524930
This item is not returnable.
View return policy
Description
VMAT2 Monoclonal Antibody for IHC (P)
VMAT2 is involved in the ATP-dependent vesicular transport of biogenic amine neurotransmitters. pumps cytosolic monoamines including dopamine, norepinephrine, serotonin, and histamine into synaptic vesicles. It is a requisite for vesicular amine storage prior to secretion via exocytosis.
Specifications
| VMAT2 | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q05940 | |
| SLC18A2 | |
| Full-length protein expressed in 293T cell transfected with human SLC18A2 expression vector. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| OTI10C11 | |
| Unconjugated | |
| SLC18A2 | |
| 1110037L13Rik; 9330105E13; MGC120477; MGC120478; MGC26538; MNAT; monoamine neurotransmitter transporter; monoamine transporter; SLC18A2; solute carrier family 18 (vesicular monoamine transporter), member 2; solute carrier family 18 (vesicular monoamine), member 2; Solute carrier family 18 A2 (vesicular monoamine transporter 2); solute carrier family 18 member 2; solute carrier family 18 member A2; solute carrier family 18, member 2; Svat; SVMT; synaptic vesicular amine transporter; VAT2; vesicle monoamine transporter type 2; vesicle monoamine/H+ antiporter; vesicular amine transporter 2; vesicular monoamine transporter 2; VMAT2; VMAT-2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6571 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction