missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VMAT2 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-68123CL1
This item is not returnable.
View return policy
Description
VMAT2 Polyclonal antibody specifically detects VMAT2 in Human,Mouse,Rabbit,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Frozen),Immunocytochemistry/Immunofluorescence
Specifications
| VMAT2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rabbit, Rat | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| MGC120477, monoamine neurotransmitter transporter, Monoamine transporter, solute carrier family 18 (vesicular monoamine), member 2, Solute carrier family 18 member 2, SVAT, SVMTMGC120478, synaptic vesicular amine transporter, VAT2MGC26538, vesicle monoamine transporter type 2, vesicle monoamine/H+ antiporter, Vesicular amine transporter 2, VMAT2 | |
| Reacts with an internal sequence of the human, mouse and rat VMAT2 protein. | |
| 0.1 mL | |
| Neuroscience | |
| 6571 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction