missing translation for 'onlineSavingsMsg'
Learn More

VMAT1/SLC18A1 Antibody (3A2-E10), Novus Biologicals™

Product Code. 18342069 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342069 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18342069 Supplier Novus Biologicals Supplier No. H00006570M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

VMAT1/SLC18A1 Monoclonal antibody specifically detects VMAT1/SLC18A1 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen VMAT1/SLC18A1
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3A2-E10
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH06317
Gene Alias solute carrier family 18 (vesicular monoamine), member 1
Host Species Mouse
Immunogen SLC18A1 (AAH06317.1, 1 a.a. ~ 493 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MLRPILDAPQRLLKEGRASRQLVLVVVFVALLLDNMLFTVVVPIVPTFLYDMEFKEVNSSLHLGHAGSSPHALASPAFSTIFSFFNNNTVAVEESVPSGIAWMNDTASTIPPPATEAISAHKNNCLQGTGFLEEETTRVGVLFASKAVMQLLVNPFVGPLTNRIGYHIPMFAGFVIMFLSTVMFAFSGTYTLLFVARTLQGIGSSFSSVAGLGMLASVYTDDHERGRAMGTALGGLALGLLVGAPFGSVMYEFVGKSAPFLILAFLALLDGALQLCILQPSKVSPESAKGTPLFMLLKDPYILVAAGLAFLPASVSYLIGTNLFGVLANKMGRWLCSLIGMLVVGTSLLCVPLAHNIFGLIGPNAGLGLAIGMVDSSMMPIMGHLVDLRHTSVYGSVYAIADVAFCMGFAIGPSTGGAIVKAIGFPWLMVITGVINIVYAPLCYYLRSPPAKEEKLAILSQDCPMETRMYATQKPTKEFPLGEDSDEEPDHEE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6570
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.