missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VLDLR Antibody (VLDLR/1337), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54553AF350
This item is not returnable.
View return policy
Description
VLDLR Monoclonal antibody specifically detects VLDLR in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| VLDLR | |
| Monoclonal | |
| Alexa Fluor 350 | |
| CARMQ1, CHRMQ1, FLJ35024, very low density lipoprotein receptor, very low-density lipoprotein receptor, VLDL receptor, VLDL-R, VLDLRCH | |
| Recombinant human VLDLR fragment from c-terminal (Uniprot: P98155) | |
| 0.1 mL | |
| Apoptosis, Cholesterol Metabolism, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| 7436 | |
| Store at 4°C in the dark. | |
| IgG1 λ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| VLDLR/1337 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction