missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
VLDLR Polyclonal specifically detects VLDLR in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | VLDLR |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Formulation | PBS |
| Gene Alias | CARMQ1, CHRMQ1, FLJ35024, very low density lipoprotein receptor, very low-density lipoprotein receptor, VLDL receptor, VLDL-R, VLDLRCH |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant VLDLR (NP_003374.3; Met 1-Ser 797). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?