missing translation for 'onlineSavingsMsg'
Learn More

VKORC1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18422086 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422086 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18422086 Supplier Novus Biologicals Supplier No. NBP281859MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VKORC1 Polyclonal antibody specifically detects VKORC1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen VKORC1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 1.1.4.1, EDTP308, FLJ00289, IMAGE3455200, MST576, phylloquinone epoxide reductase, vitamin K dependent clMST134, vitamin K epoxide reductase complex subunit 1, vitamin K epoxide reductase complex, subunit 1, Vitamin K1 2,3-epoxide reductase subunit 1, vitamin K1 epoxide reductase (warfarin-sensitive), VKCFD2, VKORMGC2694
Host Species Rabbit
Immunogen VKORC1 antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human VKORC1. The immunogen is located within the first 50 amino acids of VKORC1. Amino Acid Squence: HVKAARARDRDYRAL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79001
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.