missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Vixarelimab Recombinant Monoclonal Antibody
GREENER_CHOICE

Product Code. p-200111882
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30284064 100 μg 100µg
30283804 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30284064 Supplier Invitrogen™ Supplier No. MA559677

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Monoclonal Antibody

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Vixarelimab is a monoclonal antibody that targets and inhibits the activity of OSMRbeta (oncostatin M receptor beta), thereby reducing inflammation and pruritus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vixarelimab
Applications ELISA, Flow Cytometry, Functional Assay
Classification Recombinant Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Host Species Human
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG4SP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.