missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Vivid™ CYP2J2 Cyan Screening Kit

Product Code. 15594404
Click to view available options
Quantity:
300 Assays
Unit Size:
Pack of 300
This item is not returnable. View return policy

Product Code. 15594404

Brand: Thermo Scientific™ PV6142

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vivid™ CYP450 Screening Kits are high-throughput fluorescence-based assays for detection of enzyme-drug interactions and CYP450 inhibition.

Vivid™ CYP450 Screening Kits are high-throughput fluorescence-based assays for detection of enzyme-drug interactions and CYP450 inhibition. They provide an optimized method for studying isozyme-specific CYP450-drug interactions, metabolism, and inhibition.

Vivid™ CYP450 Screening Kits offer:

  • Easy three-step procedure, 'mix and read' format, reactions at room temperature or 37°C
  • Superior fluorescent properties and kinetics compared to conventional fluorogenic probes
  • High signal-to-background ratio, broad dynamic range
  • Compatible with multiple assay formats from 96-well to 1536-well

Simple Mix and Read Kit Format

Vivid™ CYP450 Screening Kits include Vivid™ Substrate, Vivid™ Fluorescent Standard, reaction buffer, Vivid™ Regeneration System, NADP+, and CYP450 BACULOSOMES™ Plus Reagents. The CYP450 BACULOSOMES™ Plus Reagents are microsomes prepared from insect cells expressing a human CYP450 isozyme (CYP2J2 in this case) and human cytochrome-P450 reductase. CYP450 BACULOSOMES™ Plus Reagents offer a distinct advantage over human liver microsomes in that only one CYP450 isozyme is expressed, thereby preventing metabolism by other CYP450s.

Unique Vivid™ Reagents for Bright Fluorescent Signals and Low Background

Vivid™ Substrates are blocked dyes that yield minimal fluorescence signal until cleaved or hydroxylated. Oxidation at either of two potential sites releases the highly fluorescent product (Figure 1). They have superior fluorescence, solubility, and kinetic properties compared to conventional fluorogenic probes. This results in higher sensitivity, greater signal-to-noise ratio, and better assay reproducibility. Vivid™ MOBFC Substrate is the substrate included in this particular kit. This substrate yields a product that emits a cyan fluorescence.

Flexible Assay Formats for Optimized Results

The sensitivity of the Vivid™ CYP450 assays allows detection of weak inhibitors and miniaturization to as little as 2 μl per reaction. Assays may be set up in kinetic mode or in end-point mode to facilitate multi-plate screening (Figure 2). Assays may be performed at room temperature or 37°C.

Applications:

  • High-throughput screening of enzyme-drug interactions
  • Compound profiling for drug inhibition of cytochrome P450 isozymes
  • Generation of predictive SAR models to guide compound acquisition
TRUSTED_SUSTAINABILITY

Specifications

Description Vivid™ CYP2J2 Cyan Screening Kit, 300 Assays, Cyan Color, Fluorescent Detection Method, Cytochrome P450 Druggable Target, CYP2J2 Detection Target, Human Species, Used in Cell Viability, Proliferation and Function, Pharma and Biopharma, Protein Assays
For Use With (Equipment) Microplate Reader
Product Line VIVID™
Target Cytochrome P450
For Use With (Application) Cell viability, proliferation and function, pharma and biopharma, protein assays and analysis, target-based ADME/tox assays, protein biology, cell analysis, cell metabolism, drug discovery and development, biochemical ADME assays, cytochrome p450 inhibition adme assays, protein assays and analysis, cytochrome p450 activity
Includes 1 Tube reaction buffer, 1 tube BACULOSOMES™ plus reagent, 1 tube Vivid™ regeneration system, 1 tube Vivid™ NADP+, 1 tube Vivid™ substrate and 1 tube Vivid™ fluorescent standard
Content And Storage • Reaction Buffer, 1 tube (store at room temperature)
• BACULOSOMES™ Plus Reagent, 1 tube (-80°C)
• Vivid™ Regeneration System, 1 tube (-80°C)
• Vivid™ NADP+, 1 tube (-80°C)
• Vivid™ Substrate, 1 tube (-20°C, protect from light)
• Vivid™ Fluorescent Standard, 1 tube (-20°C, protect from light)
Detection Method Fluorescence
Product Type Screening Kit
Quantity 300 Assays
Color Cyan
Shipping Condition Dry Ice
Species Human
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.