missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Vivid™ CYP2C8 Green Screening Kit

Product Code. 15371137
Change view
Click to view available options
Quantity:
300 Assays
Unit Size:
1 set
Product Code. Quantity unitSize
15371137 300 Assays 1 set
1 options
This item is not returnable. View return policy

Product Code. 15371137

Brand: Invitrogen™ PV6141

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vivid™ CYP2C8 Green Screening Kit

Vivid™ CYP450 Screening Kits are high-throughput fluorescence-based assays for detection of enzyme-drug interactions and CYP450 inhibition. They provide an optimized method for studying isozyme-specific CYP450-drug interactions, metabolism, and inhibition. Vivid™ CYP450 Screening Kits offer:

  • Easy three-step procedure, 'mix and read' format, reactions at room temperature or 37 °C
  • Superior fluorescent properties and kinetics compared to conventional fluorogenic probes
  • High signal-to-background ratio, broad dynamic range
  • Compatible with multiple assay formats from 96-well to 1536-well

    Simple Mix and Read Kit Format
    Vivid™ CYP450 Screening Kits include Vivid™ Substrate, Vivid™ Fluorescent Standard, reaction buffer, Vivid™ Regeneration System, NADP+, and CYP450 BACULOSOMES™ Plus Reagents. The CYP450 BACULOSOMES™ Plus Reagents are microsomes prepared from insect cells expressing a human CYP450 isozyme (CYP2C8 in this case) and human cytochrome-P450 reductase. CYP450 BACULOSOMES™ Plus Reagents offer a distinct advantage over human liver microsomes in that only one CYP450 isozyme is expressed, thereby preventing metabolism by other CYP450s.

    Unique Vivid™ Reagents for Bright Fluorescent Signals and Low Background
    Vivid™ Substrates are blocked dyes that yield minimal fluorescence signal until cleaved or hydroxylated. Oxidation at either of two potential sites releases the highly fluorescent product (Figure 1). They have superior fluorescence, solubility, and kinetic properties compared to conventional fluorogenic probes. This results in higher sensitivity, greater signal-to-noise ratio, and better assay reproducibility. Vivid™ DBOMF Substrate is the substrate included in this particular kit. This substrate yields a product that emits a green fluorescence.

    Flexible Assay Formats for Optimized Results
    The sensitivity of the Vivid™ CYP450 assays allows detection of weak inhibitors and miniaturization to as little as 2μl per reaction. Assays may be set up in kinetic mode or in end-point mode to facilitate multi-plate screening (Figure 2). Assays may be performed at room temperature or 37 °C.

    Applications: high-throughput screening of enzyme-drug interactions, compound profiling for drug inhibition of cytochrome P450 isozymes, generation of predictive SAR models to guide compound acquisition

    For Research Use Only. Not for any animal or human therapeutic or diagnostic use.
  • TRUSTED_SUSTAINABILITY

    Specifications

    For Use With (Equipment) Microplate Reader
    Product Line VIVID
    Target Cytochrome P450
    Content And Storage 1 tube Reaction Buffer, store at room temperature
    1 tube BACULOSOMES™ Plus Reagent, store at -80 °C
    1 tube Vivid™ Regeneration System, store at -80 °C
    1 tube Vivid™ NADP+, store at -80 °C
    1 tube Vivid™ Substrate and 1 tube Vivid™ Fluorescent Standard, store at -20 °C, protected from light
    Detection Method Fluorescence
    Product Type Screening Kit
    Quantity 300 Assays
    Format Kit
    Color Green
    Shipping Condition Dry Ice
    Species Human
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.