Learn More
Thermo Scientific™ Vivid™ BOMR Substrate
Description
Vivid™ Substrates are blocked dyes that yield a minimal fluorescence signal until oxidation occurs. Cytochrome P450 metabolism at either of two potential oxidation sites releases the highly fluorescent product. They have superior fluorescence, solubility, and kinetic properties compared to conventional fluorogenic probes. This results in higher sensitivity, greater signal-to-noise ratio, and better assay reproducibility. When used in combination with CYP450 BACULOSOMES™ Plus Reagents as part of the Vivid™ CYP450 Screening Kits, the Vivid™ Substrates enable rapid, robust detection of cytochrome P450 inhibitors. This particular substrate (Vivid ™ BOMR Substrate) yields a product that emits a red fluorescence.
More About Vivid™ CYP450 Screening Kits
Vivid™ CYP450 Screening Kits are high-throughput fluorescence-based assays for detection of enzyme-drug interactions and CYP450 inhibition. Vivid™ CYP450 Screening Kits include Vivid™ Substrate, Vivid™ Fluorescent Standard, reaction buffer, Vivid™ Regeneration System, NADP+, and CYP450 BACULOSOMES™ Plus Reagent. CYP450 BACULOSOMES™ Plus Reagents are microsomes prepared from insect cells expressing a human CYP450 isozyme and human cytochrome P450 reductase. CYP450 BACULOSOMES™ Plus Reagents offer a distinct advantage over human liver microsomes in that only one CYP450 isozyme is expressed, thereby preventing metabolism by other CYP450s.
Applications
- High-throughput screening of enzyme-drug interactions
- Compound profiling for drug inhibition of cytochrome P450 isozymes
- Generation of predictive SAR models to guide compound acquisition
Specifications
Specifications
| Chromagen Quantity | 1mg |
| Content And Storage | Vivid™ CYP450 Screening kits include: • Vivid™ CYP450 Fluorogenic substrates (also available separately) • BACULOSOMES™ CYP450 reagents • Fluorescent standards • NADPH-regeneration system Store reaction buffers at +4°C or room temperature. Store BACULOSOMES™, the NADPH-regeneration system, and NADP+ at -80°C. Store the Vivid™ substrates and standards at +4°C, protected from light. (For long-term storage, resuspend in organic solvent and keep at -20°C.) |
| Detection Method | Fluorescence |
| For Use With (Application) | Cell viability, proliferation and function, pharma and biopharma, protein assays and analysis, target-based ADME/Tox assays |
| Product Line | VIVID |
| Quantity | 1 mg |
| Substrate | BOMR |
| Substrate Properties | Chemical Substrate |
| Substrate Type | Cytochrome P450 Substrate |
| Item Description | Vivid™ BOMR Substrate, 1mg, Red Fluorescent, Dried under Vacuum from Acetonitrile, Blocked Dyes that Yield a Minimal Fluorescence Signal until Oxidation Occurs |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.