Learn More
Sartorius™ Vivacon™ Polycarbonate/Polypropylene Centrifugal Unit
Brand: Sartorius™ VN01H42ETO
This item is not returnable.
View return policy
Description
Specifications
| PCR | |
| 100,000 Da | |
| Polycarbonate, Polypropylene | |
| DNA | |
| 0.1 to 0.5 mL |
| Polycarbonate, Polypropylene | |
| Concentration, Desalting, buffer exchange | |
| 100/Pk. | |
| Vivacon 500 Centrifugal Unit |