missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Vivacon 500 125kDa
Shop All Sartorius Products

Click to view available options
Quantity:
100 Pk.
25 Pk.
Unit Size:
Pack of 100
Pack of 25
Description
- The product of choice for protein fractionation applications.
- The use of Hydrosart™ membranes results in high recoveries from dilute samples.
- Complete and highly reproducible recovery of the concentrate.
- Reverse spinning step capacity.
- Concentration of up to 0.5 mL genomic DNA or peptide samples.
- Low binding material.
- MWCO printed on device.
- Selected cut offs available sterile and DNA-free for things like forensic applications.
- qPCR assay over 55 cycles with a verified limit of detection <2 pg
Specifications
Specifications
| Color | Transparent |
| Volume (Metric) | 0.5 mL |
| Quantity | 25 Pk. |
| Product Line | Centrifugal ultrafiltration device |
| Sterility | Sterile |
| Material | Polypropylene |
| Closure Type | Flat Cap (Attached) |
| Autoclavable | Not Autoclavable |
| Closure Included | Yes |
| Disposable | Single-use |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction