missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Vivacon 2 125kDa
Shop All Sartorius Products

Click to view available options
Quantity:
100/Pk.
25/Pk.
Unit Size:
Pack of 100
Pack of 25
Description
- The product of choice for protein fractionation applications.
- The use of Hydrosart™ membranes results in high recoveries from dilute samples.
- Complete and highly reproducible recovery of the concentrate.
- Reverse spinning step capacity.
- Concentration of up to 0.5 mL genomic DNA or peptide samples.
- Low binding material.
- MWCO printed on device.
- Selected cut offs available sterile and DNA-free for things like forensic applications.
- qPCR assay over 55 cycles with a verified limit of detection < 2 pg
Specifications
Specifications
| Description | MWCO: 125,000 Daltons |
| Volume (Metric) | 0.1 to 2.0 mL, 55 μL (Dead Stop Volume) |
| Quantity | 100/Pk. |
| Product Line | Centrifugal ultrafiltration device |
| Material | Polycarbonate Concentrator Body, Polypropylene Filtrate Vessel, Cellulose Acetate Membrane |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction