Learn More
STEMCELL Technologies Vitronectin XF™ Kit with GCDR
Product Code. 17159241
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. Quantity unitSize
17159241 1 Kit 1 set
1 options
Product Code. 17159241 Supplier STEMCELL Technologies Supplier No. 07190
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For maintenance of human ES/iPS cells in combination with TeSR™-E8™, mTeSR™1, or TeSR™2
  • Vitronectin XF™, developed and manufactured by Nucleus Biologics, is an effective alternative to Corning™ Matrigel™
  • Vitronectin XF™ is a defined, xeno-free cell culture matrix that supports the growth and differentiation of human pluripotent stem cells
  • When used with mTeSR™1, mTeSR™ Plus, TeSR™-E8™, or TeSR™2, Vitronectin XF™ provides a defined culture system for the maintenance of human embryonic stem (ES) cells and human induced pluripotent stem (iPS) cells under feeder-free conditions
  • This system allows complete control over the culture environment, resulting in more consistent cell populations and reproducible results in downstream applications
  • Note: Catalog #07180 is the matrix only and requires additional reagents (Catalog #07183, 07174 or 05872, and 27147) available for purchase separately or in a kit with Vitronectin XF™ (Catalog #07190 or 07191)
Specifications
Cell Type Endoderm, PSC-Derived, Pluripotent Stem Cells
Quantity 1 Kit