missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Vitronectin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ 720229
This item is not returnable.
View return policy
Description
This antibody is predicted to react with Monkey, Dog, Mouse.
VTN is a member of the pexin family. This protein is found in serum and tissues and promotes cell adhesion and spreading, inhibits the membrane-damaging effect of the terminal cytolytic complement pathway, and binds to several serpin serine protease inhibitors. The protein is a secreted protein and exists in either a single chain form or a clipped, two chain form held together by a disulfide bond.
Specifications
| Vitronectin | |
| Polyclonal | |
| Unconjugated | |
| VTN | |
| Aa1018; AI256434; complement S-protein; epibolin; extracellular matrix protein; glycoprotein 66; serum spreading factor; serum-spreading factor; somatomedin B; Somatomedin-B; S-protein; V75; Vitronectin; vitronectin (serum spreading factor, somatomedin B, complement S-protein); Vitronectin V10 subunit; Vitronectin V65 subunit; VN; VNT; VTN | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7448 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.4 | |
| P04004 | |
| VTN | |
| Peptides corresponding to human VTN (aa367-384, 107-120). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction