Learn More
Vitronectin Antibody (A18), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-23571DL594
Product Code. 18454696
  • £561.00 / 0.25mL
Description
Vitronectin Monoclonal antibody specifically detects Vitronectin in Bovine, Equine, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vitronectin
Monoclonal
DyLight 594
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Bovine, Equine, Rabbit
Purified
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
A18
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
complement S-protein, epibolin, serum spreading factor, Serum-spreading factor, somatomedin B, S-protein, V75, vitronectin, vitronectin (serum spreading factor, somatomedin B, complement S-protein), VN, VNT
Lysed bovine corneal endothelial cells and extracellular matrix
0.25 mL
Immunology, Innate Immunity
7448
Store at 4°C in the dark.
IgG1 κ