missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitronectin Antibody (019), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90648B
This item is not returnable.
View return policy
Description
Vitronectin Monoclonal specifically detects Vitronectin in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Vitronectin | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 19 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| complement S-protein, epibolin, serum spreading factor, Serum-spreading factor, somatomedin B, S-protein, V75, vitronectin, vitronectin (serum spreading factor, somatomedin B, complement S-protein), VN, VNT | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Vitronectin (NP_035837.1; Met1-Lys478). | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 7448 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction