missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitrin Polyclonal Antibody
Vitrin Polyclonal Antibody
Brand: Invitrogen PA524009
This item is not returnable.
View return policy
Description
Vitrin Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
| Vitrin | |
| Polyclonal | |
| Unconjugated | |
| VIT | |
| UNQ647/PRO1277; VIT; Vitrin | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 5212 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6UXI7 | |
| VIT | |
| KLH conjugated synthetic peptide between 567-595 amino acids from the C-terminal region of human VITRN. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction