missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Vito S3 Shoe

Product Code. 17592436
Change view
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. Size unitSize
17592436 41 1 pair
17572436 39 1 pair
17582436 40 1 pair
17502446 42 1 pair
17512446 43 1 pair
17522446 44 1 pair
17532446 45 1 pair
17542446 46 1 pair
17552446 47 1 pair
17562446 48 1 pair
10 options
This item is not returnable. View return policy

Product Code. 17592436

Brand: Emma Safety Footwear MM11400041

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Maintain that sharp look, even when wearing your safety gear with these Emma Vito safety shoes.

  • Smart work shoes with water-resistant full-grain and nubuck leather uppers
  • Hydro-Tec antibacterial and Wellness leather lining
  • Antibacterial inlays with leather top layers
  • Steel toecaps and aramid fibre penetration-resistant midsoles
  • Polyurethane (PU) comfort midsoles with thermoplastic polyurethane (TPU) outsoles
  • Regular fit
  • Worn shoes can be returned to the manufacturer for recycling (postage provided); keep receipt and contact us for more details when the time comes
  • Outsoles are resistant to fuel oil
  • Excellent slip resistance
  • Energy-absorbing heels for comfort and support
  • Antistatic

Specifications

Certifications/Compliance ISO 20345:2011 S3 FO
Closure Type Laces
Color Black
For Use With (Application) Government, Local Authority, Oil and Gas
Material Leather
Size 41
Toe Material Steel
Fit Extra-wide Width
Ankle Style Low-cut
Product Type Safety Shoe
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.