missing translation for 'onlineSavingsMsg'
Learn More

Vitamin K4, MedChemExpress

Product Code. 16491456
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
1 g
Unit Size:
1g
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491456 200 mg 200mg
16432349 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16491456 Supplier Medchem Express Supplier No. HYB1508
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Vitamin K4 is a chemically synthesized Vitamin K which plays an important role in the normal blood coagulation system.

Vitamin K4 is a chemically synthesized Vitamin K which plays an important role in the normal blood coagulation system.

Specifications

Chemical Name or Material Vitamin K4
CAS 573-20-6
Color White
Purity Grade Notes Research
Molecular Formula C15H14O4
Quantity 200 mg
Synonym acetomenaphthone
Solubility Information DMSO : 100 mg/mL (387.19 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (2.59 mM; Need ultrasonic)
SMILES CC1=CC(OC(C)=O)=C2C=CC=CC2=C1OC(C)=O
Molecular Weight (g/mol) 258.27
Formula Weight 258.27
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.