missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Vitamin K3 Monoclonal Antibody (5H9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549644
This item is not returnable.
View return policy
Description
Antibody tested in indirect ELISA.
Vitamin K3, also known as menadione, is a synthetic form of vitamin K that is used as a nutritional supplement. It is commonly used in animal feed to prevent vitamin K deficiency and improve overall health and growth. Vitamin K3 is also used in the production of various food and pharmaceutical products. It plays a critical role in blood clotting and bone metabolism, and it has been shown to have antioxidant and anti-inflammatory properties. However, excessive intake of vitamin K3 can be toxic and can cause liver damage.
Specifications
| Vitamin K3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry, Lateral Flow | |
| 5H9 | |
| Unconjugated | |
| menadione | |
| Conjugate of VK3 hapten and KLH. | |
| 200 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction