missing translation for 'onlineSavingsMsg'
Learn More

Vitamin K1, MedChemExpress

Product Code. 16481478
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481478 5 g 5g
16432549 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481478 Supplier Medchem Express Supplier No. HYN0684
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Vitamin K1 a naturally occurring vitamin required for blood coagulation and bone and vascular metabolism.

Vitamin K1 a naturally occurring vitamin required for blood coagulation and bone and vascular metabolism.

Specifications

Chemical Name or Material Vitamin K1
CAS 84-80-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C31H46O2
Quantity 5 g
Synonym Phylloquinone Phytomenadione
Solubility Information Ethanol : ≥ 50 mg/mL (110.94 mM) ∣DMSO : 5.6 mg/mL (12.43 mM; Need ultrasonic and warming)
SMILES O=C1C(C)=C(C/C=C(C)/CCC[C@H](C)CCC[C@H](C)CCCC(C)C)C(C2=C1C=CC=C2)=O
Molecular Weight (g/mol) 450.7
Formula Weight 450.7
Percent Purity 98.0%
Grade Research
Physical Form Liquid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.