missing translation for 'onlineSavingsMsg'
Learn More

Vitamin K1 2,3-Epoxide (Mixture of Diastereomers), TRC

Product Code. 30010598
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010598 50 mg 50mg
30005529 250 mg 250mg
30007638 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010598 Supplier Toronto Research Chemicals Supplier No. TRCV68001350MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25486-55-9
Molecular Formula C31 H46 O3
Molecular Weight (g/mol) 466.7
Synonym Naphth[2,3-b]oxirene-2,7-dione, 1a,7a-dihydro-1a-methyl-7a-(3,7,11,15-tetramethyl-2-hexadecen-1-yl)-, 1,4-Naphthoquinone, 2,3-epoxy-2,3-dihydro-2-methyl-3-phytyl- (6CI,7CI,8CI), Naphth[2,3-b]oxirene-2,7-dione, 1a,7a-dihydro-1a-methyl-7a-(3,7,11,15-tetramethyl-2-hexadecenyl)- (9CI), 1a,7a-Dihydro-1a-methyl-7a-(3,7,11,15-tetramethyl-2-hexadecen-1-yl)naphth[2,3-b]oxirene-2,7-dione, 2,3-Epoxyphylloquinone, Phylloquinone oxide, Phylloquinone, epoxide, Phylloquinone-2,3-epoxide, Vitamin K 2,3-epoxide, Vitamin K epoxide, Vitamin K oxide, Vitamin K1 2,3-epoxide, Vitamin K1 oxide, Vitamin K1, epoxide
IUPAC Name 7a-methyl-1a-[(E)-3,7,11,15-tetramethylhexadec-2-enyl]naphtho[2,3-b]oxirene-2,7-dione
SMILES CC(C)CCCC(C)CCCC(C)CCC\C(=C\CC12OC1(C)C(=O)c3ccccc3C2=O)\C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C31H46O3/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-17-25(5)20-21-31-29(33)27-19-8-7-18-26(27)28(32)30(31,6)34-31/h7-8,18-20,22-24H,9-17,21H2,1-6H3/b25-20+
Formula Weight 466.3447
Percent Purity >95%
Chemical Name or Material Vitamin K1 2,3-Epoxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.