missing translation for 'onlineSavingsMsg'
Learn More

Vitamin K, MedChemExpress

Product Code. 16414657
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16414657 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16414657 Supplier Medchem Express Supplier No. HYB2172

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vitamin K, the blood-clotting vitamin, is important for the function of numerous proteins within the body, such as the coagulation factors, osteocalcin and matrix-Gla protein.

Vitamin K, the blood-clotting vitamin, is important for the function of numerous proteins within the body, such as the coagulation factors, osteocalcin and matrix-Gla protein.

Specifications

Chemical Name or Material Vitamin K
CAS 12001-79-5
Color Yellow
Purity Grade Notes Research
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (Need ultrasonic)
SMILES [Vitamin K]
Percent Purity 98.0%
Grade Research
Physical Form Liquid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.