missing translation for 'onlineSavingsMsg'
Learn More

α-Vitamin E, MedChemExpress

Product Code. 16422549
Encompass_Preferred
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
This item is not returnable. View return policy

Product Code. 16422549

Brand: Medchem Express HYN0683

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress α-Vitamin E ((+)-α-Tocopherol), a naturally occurring vitamin E form, is a potent antioxidant.

α-Vitamin E ((+)-α-Tocopherol), a naturally occurring vitamin E form, is a potent antioxidant.

Specifications

Chemical Name or Material α-Vitamin E
CAS 59-02-9
Color Colorless
Purity Grade Notes Research
Molecular Formula C29H50O2
Quantity 1 g
Synonym (+)-α-Tocopherol D-α-Tocopherol
Solubility Information DMSO : 50 mg/mL (116.09 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES OC1=C(C)C(C)=C2C(CC[C@](CCC[C@H](C)CCC[C@H](C)CCCC(C)C)(C)O2)=C1C
Molecular Weight (g/mol) 430.71
Formula Weight 430.71
Percent Purity 98.0%
Grade Research
Physical Form Viscous Liquid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.