missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Vitamin D3 Receptor Recombinant Protein

Product Code. 15420714
Click to view available options
Quantity:
30 μg
Unit Size:
30µg
This item is not returnable. View return policy

Product Code. 15420714

Brand: Invitrogen™ A15670

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This full-length human Vitamin D3 Receptor is purified from recombinant baculovirus-infected insect cells and is soluble and functionally active with an observed Kd for [3H]-1a, 25-dihydroxyvitamin D3 of 2 nM. Vitamin D3 Receptor human recombinant protein is:, Flexible-use in your in-house assay or as part of our PolarScreenTM Competitor assays for a complete assay solution, Ready to use-active and native (untagged), Convenient-available in bulk sizes Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond. Protein Form: Recombinant, Full Length

Steroid receptors are ligand-dependent intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate hormone. The 1,25-dihydroxy-vitamin D3 receptor (VDR) belongs to the superfamily of steroid hormone receptors which includes estrogen, progesterone, glucocorticoid, androgen, and thyroid hormone receptors. Vitamin D modulates calcium and phosphorus homeostasis, bone remodeling, cell growth regulation, and differentiation. Studies have found VDR in the intestine, bone, kidney, epidermis, and cells of the endocrine immune system. Repression of T-cell proliferation and interleukin-2 (IL-2) gene expression, for example, occurs when VDR binds within the IL-2 enhancer. Formation of VDR/retinoic X receptor (RXR) heterodimers in the presence of intracellular 1,25(OH)2D3 has been shown to interfere with assembly of nuclear factor of activated T-cells (NFATp)/Fos/Jun/DNA complex and subsequent IL-2 gene transcription. The corresponding gene for the vitamin D receptor is NR1I1.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P11473
Concentration See Label
For Use With (Application) Kinase Assay
Formulation proprietary buffer with no preservative
Gene ID (Entrez) 7421
Molecular Weight (g/mol) 48 kDa
Name Human Vitamin D3 Receptor
Quantity 30 μg
Regulatory Status RUO - research use only
Gene Alias 1,25-dihydroxyvitamin D3 receptor; HGNC:12679; I79_025795; Nr1i1; Nuclear receptor subfamily 1 group I member 1; PPP1R163; protein phosphatase 1, regulatory subunit 163; vdr; vdr.S; vdr-A; vitamin D (1,25- dihydroxyvitamin D3) receptor; vitamin D (1,25- dihydroxyvitamin D3) receptor S homeolog; vitamin D (1,25-dihydroxyvitamin D3) receptor; vitamin D nuclear receptor variant 1; vitamin D receptor; vitamin D receptor protein; vitamin D receptor S homeolog; Vitamin D3 receptor; XELAEV_18015993mg; xVDR
Common Name VDR
Gene Symbol VDR
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Baculovirus
Form Liquid
Purity or Quality Grade 50% as determined by a SDS-PAGE gel stained with SimplyBlueTM SafeStain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.