Learn More
Invitrogen™ Vitamin D3 Receptor Human Protein, Ligand Binding Domain
Vitamin D3 Receptor, Ligand Binding Domain (VDR-LBD) is a 65.50 kDa recombinant human protein (amino acids 93-427) expressed as a GST-tagged fusion protein in baculovirus-infected insect cells.
Brand: Invitrogen™ PV4552
Code : 50
Additional Details : Weight : 0.50000kg
Description
VDR-LBD was expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator displacement studies.
Specifications
Nuclear Receptor Assay | |
50μg | |
RUO - research use only | |
Vitamin D3 Receptor | |
Recombinant |
7421 | |
Store in ultra-cold freezer (-68 to -85°C). | |
Ligand Binding Domain | |
Human | |
Hormone Receptors |