missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitamin D2 Sulfate Sodium Salt, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Vitamin D2 Sulfate Sodium Salt |
| CAS | 85704-61-6 |
| Molecular Formula | C28H43NaO4S |
| Quantity | 10 mg |
| Synonym | (3β,5Z,7E,22E)-9,10-Secoergosta-5,7,10(19),22-tetraen-3-ol Hydrogen Sulfate, Ergocalciferol Hydrogen Sulfate, Vitamin D2 3β-Sulfate, Vitamin D2 Sulfate, (3beta,5Z,7E,22E)-9,10-Secoergosta-5,7,10(19),22-tetraen-3-ol Hydrogen Sulfate, Vitamin D2 3beta-Sulfate |
| InChI Formula | InChI=1S/C28H44O4S.Na/c1-19(2)20(3)9-10-22(5)26-15-16-27-23(8-7-17-28(26,27)6)12-13-24-18-25(14-11-21(24)4)32-33(29,30)31;/h9-10,12-13,19-20,22,25-27H,4,7-8,11,14-18H2,1-3,5-6H3,(H,29,30,31);/q;+1/p-1/b10-9+,23-12+,24-13-;/t20-,22+,25-,26+,27-,28+;/m0./s1 |
| SMILES | [H][C@@]1(CC[C@]2([H])[C@@]1(CCC/C2=C\C=C3C[C@H](CCC\3=C)OS(=O)(O[Na])=O)C)[C@@H](/C=C/[C@@H](C(C)C)C)C |
| IUPAC Name | sodium (S,Z)-3-(2-((1R,3aS,7aR,E)-1-((2R,5R,E)-5,6-dimethylhept-3-en-2-yl)-7a-methyloctahydro-4H-inden-4-ylidene)ethylidene)-4-methylenecyclohexyl sulfate |
| Molecular Weight (g/mol) | 498.69 |
| Formula Weight | 498.28 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction