missing translation for 'onlineSavingsMsg'
Learn More

Vitamin D BP Antibody (VDBP/4482), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18319066 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18319066 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18319066 Supplier Novus Biologicals Supplier No. NBP314006AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Vitamin D BP Monoclonal antibody specifically detects Vitamin D BP in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vitamin D BP
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone VDBP/4482
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DBP/GC, DBPVDBG, EC 6.3.1.5, Gc-globulin, GRD3, Group-specific component, group-specific component (vitamin D binding protein), hDBP, VDB, VDBP, vitamin D-binding alpha-globulin, vitamin D-binding protein
Host Species Mouse
Immunogen Recombinant fragment (around aa 35-175) of human Vitamin D BP protein (exact sequence is proprietary) (Uniprot: P02774)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2638
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.