missing translation for 'onlineSavingsMsg'
Learn More
Learn More
vitamin D binding protein Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16922-1-AP-20UL
This item is not returnable.
View return policy
Description
GC (vitamin D-binding protein) is involved in vitamin D transport/storage, scavenging of extracellular G-actin, enhancement of the chemotactic activity of C5 alpha for neutrophils in inflammation and macrophage activation.Specifications
| vitamin D binding protein | |
| Polyclonal | |
| Unconjugated | |
| GC | |
| DBP, DBP/GC, GC, Gc globulin, Group specific component, VDB, VDBG, VDBP, Vitamin D binding protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14473, 24384, 2638 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P02774, P04276, P21614 | |
| Gc | |
| vitamin D binding protein Fusion Protein Ag10286 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction