missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitamin B2 (Riboflavin) 13C4 ,15N2, TRC
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
Specifications
Specifications
| Chemical Name or Material | Riboflavin-2,4,4a,10a-13C4-1,3-15N2 |
| CAS | 1217461-14-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | 13C4 C13 H20 15N2 N2 O6 |
| Quantity | 0.5 mg |
| Synonym | Riboflavin-2,4,4a,10a-13C4-1,3-15N2, Riboflavin-13C4,15N2, Vitamin B2-13C4,15N2, 1-Deoxy-1-(3,4-dihydro-7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10(2H)-yl-2,4,4a,10a-13C4-1,3-15N2)-D-ribitol |
| InChI Formula | InChI=1S/C17H20N4O6/c1-7-3-9-10(4-8(7)2)21(5-11(23)14(25)12(24)6-22)15-13(18-9)16(26)20-17(27)19-15/h3-4,11-12,14,22-25H,5-6H2,1-2H3,(H,20,26,27)/t11-,12+,14-/m0/s1/i13+1,15+1,16+1,17+1,19+1,20+1 |
| SMILES | Cc1cc2N=[13C]3[13C](=O)[15NH][13C](=O)[15N]=[13C]3N(C[C@H](O)[C@H](O)[C@H](O)CO)c2cc1C |
| IUPAC Name | 7,8-dimethyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]benzo[g]pteridine-2,4-dione |
| Molecular Weight (g/mol) | 382.32 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction