missing translation for 'onlineSavingsMsg'
Learn More

Vitamin B12, 98+% (dry wt basis)

Product Code. 11452697
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452697 5 g 5g
11442697 1 g 1g
11462697 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452697 Supplier Thermo Scientific Alfa Aesar Supplier No. A14894.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a nutritional supplement and stabilizer in foods. | CAS: 68-19-9 | C63H88CoN14O14P | 1355.39 g/mol

Vitamin B12 is used as a nutritional supplement and stabilizer in foods. It is also used as a medicine and animal feed supplement. It plays an important role in the synthesis of myelin. It is also used as an antipernicious anemia factor. Further, it is used to treat cyanide poisoning and hereditary deficiency of transcobalamin II.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-19-9
Molecular Formula C63H89CoN14O14P
Molecular Weight (g/mol) 1356.396
MDL Number MFCD00151092
InChI Key AGVAZMGAQJOSFJ-WZHZPDAFSA-M
Synonym vitamin b12
PubChem CID 129893524
IUPAC Name cobalt(2+);[(2R,3S,4R,5S)-5-(5,6-dimethylbenzimidazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl] [(2R)-1-[3-[(1R,2R,3R,5Z,7S,10Z,12S,13S,15Z,17S,18S,19R)-2,13,18-tris(2-amino-2-oxoethyl)-7,12,17-tris(3-amino-3-oxopropyl)-3,5,8,8,13,15,18,19-octamethyl-
SMILES CC1=CC2=C(C=C1C)N(C=N2)C3C(C(C(O3)CO)OP(=O)(O)OC(C)CNC(=O)CCC4(C(C5C6(C(C(C(=C(C7=NC(=CC8=NC(=C(C4=N5)C)C(C8(C)C)CCC(=O)N)C(C7(C)CC(=O)N)CCC(=O)N)C)[N-]6)CCC(=O)N)(C)CC(=O)N)C)CC(=O)N)C)O.[C-]#N.[Co+2]

Specifications

Packaging Glass bottle
Quantity 5 g
Beilstein 4122889
Sensitivity Hygroscopic
Solubility Information Soluble in water,dimethyl sulfoxide and ethanol. Insoluble in chloroform and acetone.
Formula Weight 1355.38
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material Vitamin B12

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.