missing translation for 'onlineSavingsMsg'
Learn More

Vitamin A Acetate, TRC

Product Code. 30010201
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Unit Size:
100g
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010201 100 g 100g
30009219 5 g 5g
30010481 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010201 Supplier Toronto Research Chemicals Supplier No. TRCV676000100G
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-47-9
Molecular Formula C22 H32 O2
Molecular Weight (g/mol) 328.49
Synonym Retinol, acetate, all-trans- (8CI), Arovit, Myvak, Myvax, NSC 122045, NSC 122760, Retinol acetate, Retinyl acetate, Ro 1-5275, Rovimix A 1000, Vitamin A acetate, Vitamin A1 acetate, all-trans-Retinol acetate, all-trans-Retinyl acetate, all-trans-Vitamin A acetate, trans-Retinol acetate, trans-Retinyl acetate
IUPAC Name [(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate
SMILES CC(=O)OC\C=C(/C)\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C

Specifications

Quantity 100 g
InChI Formula InChI=1S/C22H32O2/c1-17(9-7-10-18(2)14-16-24-20(4)23)12-13-21-19(3)11-8-15-22(21,5)6/h7,9-10,12-14H,8,11,15-16H2,1-6H3/b10-7+,13-12+,17-9+,18-14+
Formula Weight 328.2402
Percent Purity >95%
Chemical Name or Material Vitamin A acetate (Retinol acetate)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.