missing translation for 'onlineSavingsMsg'
Learn More

Vistusertib, MedChemExpress

Product Code. p-200099403
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16476686

Brand: Medchem Express HY15247

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vistusertib (AZD2014) is an ATP competitive mTOR inhibitor with an IC50 of 2.81 nM. AZD2014 inhibits both mTORC1 and mTORC2 complexes.

Vistusertib (AZD2014) is an ATP competitive mTOR inhibitor with an IC50 of 2.81 nM. AZD2014 inhibits both mTORC1 and mTORC2 complexes.

Specifications

Chemical Name or Material Vistusertib
CAS 1009298-59-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C25H30N6O3
Quantity 100 mg
Synonym AZD2014
Solubility Information DMSO : ≥ 50 mg/mL (108.10 mM)
SMILES O=C(NC)C1=CC=CC(C2=CC=C3C(N=C(N4[C@@H](C)COCC4)N=C3N5[C@@H](C)COCC5)=N2)=C1
Molecular Weight (g/mol) 462.54
Formula Weight 462.54
Percent Purity 95.66%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.