missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies Ovation™ Pipet Power Supply

For all countries

Brand:  VistaLab™ Technologies 9060-9005

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11976895

  • £44.60 / Each
Estimated Shipment: 25-06-2024
to see stock.
Add to basket

Description

Description

  • 90–264VAC, 47–63Hz
  • Available with or without pipet connections

Specifications

Specifications

Non-sterile
Without connections
3 in.
5 in.
90/264 V
Ovation™ Electronic Single Channel, Macro and Multichannel Models
1 Ea.
3 in.
90/264 V 47/63 Hz
47/63 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers