missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies MLA™ Brand Precision Pipetters
GREENER_CHOICE

Product Code. 11915782
Click to view available options
Volume (Metric):
10 μL
20 μL
25 μL
50 μL
100 μL
150 μL
200 μL
250 μL
300 μL
500 μL
1000 μL
Color:
Black
Blue
Brown
Gray
Green
Orange
Purple
Red
Silver
White
Yellow
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11915782

Brand: VistaLab™ Technologies 1021

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ten fixed-volume models from 10 to 1000μL

  • 'To deliver' capability ensures volume accuracy over a wide range of viscosities
  • Ideal for repetitive delivery of reagents when high precision and accuracy are required
  • Sleeves are color coded for easy volume identification
  • Capacity engraved on bonnet

Order Info

Additional sizes available. Contact your Fisher Scientific Customer Service Representative for more information.

Specifications

Accuracy ±5%
Autoclavable Not Autoclavable
Color Orange
Disposable Reusable
For Use With Micropipette Tip
Material Aluminum, Stainless Steel
Model F1
Number of Channels 1
Sterility Non-sterile
Type Precision Pipette
Volume (Metric) 10 μL
Includes Nozzle Insert, Calibration Key
Quantity 1 Ea.
Warranty Limited Lifetime
Length (English) 9 in.
Width (English) 2 in.
Height (English) 1.5 in.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.