missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies Pipette Tips: Low Retention
GREENER_CHOICE

Product Code. 11915992
Click to view available options
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
Volume (Metric):
2 μL
25 μL
250 μL
1250 μL
Unit Size:
Pack of 960
This item is not returnable. View return policy

Product Code. 11915992

Brand: VistaLab™ Technologies 40701333LR

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with Ovation and MLA Brand pipetters

  • Sterile, Low Retention tips packaged in VistaRak: 5 racks of 192 tips each
  • Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, etc.
  • Use VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation pipettes
  • All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time

Specifications

Description Low Retention
Volume (Metric) 2 μL
Filtered Filtered
Sterility Sterile
Color Clear
Rack Color Blue
Autoclavable Not Autoclavable
Length (English) 1.7 in.
Thickness 5 in.
For Use With (Application) Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, TRIS, BSA and Tween, PCR or other biological applications
Sterilization Method e-beam
Format Rack
Tip Style Low Retention, Straight
Packaging VistaRak: 5 racks
Quantity 192 Tips/Rack
Product Type Pipette Tips
Color Code Blue
Disposable Single-use
For Use With (Equipment) Ovation
Graduated No
Material Polypropylene, Polyethylene (Filter)
Shape Cone
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.