missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VISTA Mouse anti-Human , BV421, Clone: MIH65.rMAb (also known as MIH65), BD Biosciences™
Mouse Monoclonal Antibody
£197.74 - £779.28
Specifications
| Antigen | VISTA |
|---|---|
| Clone | MIH65.rMAb (also known as MIH65) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Violet 421 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17287574
|
BD Biosciences
566751 |
25 Tests |
£207.00 £197.74 / 25 tests Save £9.26 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17277574
|
BD Biosciences
566750 |
100 Tests |
£788.00 £779.28 / 100 tests Save £8.72 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VISTA Monoclonal antibody specifically detects VISTA in Human samples. It is validated for Flow CytometrySpecifications
| VISTA | |
| Flow Cytometry | |
| Brilliant Violet 421 | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human VISTA Transfected Cell Line | |
| Primary |
| MIH65.rMAb (also known as MIH65) | |
| Monoclonal | |
| Mouse | |
| Human | |
| VSIR; B7-H5; B7H5; C10orf54; DD1alpha; GI24; SISP1 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV421 under optimum conditions, and unconjugated antibody and free BD Horizon BV421 were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title