missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VISTA Monoclonal Antibody (MIH64), Super Bright™ 600, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
£142.00 - £346.00
Specifications
| Antigen | VISTA |
|---|---|
| Clone | MIH64 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
Rat Monoclonal Antibody
VISTA (V-domain Ig containing suppressor of T-cell activation), also known as B7-H5 or PD-1H, is a product of the VSIR gene. Although VISTA bears sequence homology to PD-L1 it has a different expression pattern. In both mouse and humans, VISTA is expressed predominantly on hematopoietic cells with the highest densities on myeloid cells, and lower levels on T cells. It is usually absent on B cells. VISTA expressed by antigen presenting cells has been shown to directly suppress proliferation and cytokine production of CD4 and CD8 T cells, making it a target for cancer immunotherapy. The reported ligands of VISTA include CD28H, VSIG-3 and VSIG-8.Specifications
| VISTA | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| 74048 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| MIH64 | |
| Flow Cytometry | |
| Super Bright 600 | |
| Rat | |
| Mouse | |
| Q9D659 | |
| VSIR | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! | |
| VSIR |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title