missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VISTA Monoclonal Antibody (MIH64), Brilliant Ultra Violet™ 737, eBioscience™, Invitrogen™
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ 367-1083-82

 View more versions of this product

Product Code. 17433382

  • £353.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This MIH64 monoclonal antibody recognizes mouse VISTA also known as B7-H5 or PD-H1. This MIH64 antibody will work in flow cytometry on both native and paraformaldehyde-fixed cells. Applications Reported: This MIH64 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MIH64 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 1.0 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Brilliant Ultraviolet 737 is a tandem dye that emits at 732 nm and is intended for use on cytometers equipped with an ultraviolet (355 nm) laser.

VISTA (V-domain Ig containing suppressor of T-cell activation), also known as B7-H5 or PD-1H, is a product of the VSIR gene. Although VISTA bears sequence homology to PD-L1 it has a different expression pattern. In both mouse and humans, VISTA is expressed predominantly on hematopoietic cells with the highest densities on myeloid cells, and lower levels on T cells. It is usually absent on B cells. VISTA expressed by antigen presenting cells has been shown to directly suppress proliferation and cytokine production of CD4 and CD8 T cells, making it a target for cancer immunotherapy. The reported ligands of VISTA include CD28H, VSIG-3 and VSIG-8.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

VISTA
Monoclonal
0.2 mg/mL
PBS with BSA and 0.09% sodium azide; pH 7.2
Q9D659
VSIR
Affinity chromatography
RUO
74048
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
MIH64
Brilliant Ultraviolet 737
VSIR
4632428N05Rik; B7H5; B7-H5; C10orf54; C1H10orf54; C28H10orf54; C4H10orf54; chromosome 10 open reading frame 54; DD1alpha; Death Domain1alpha; Dies1; differentiation of ESCs 1; GI24; PD-1H; PDCD1 homolog; PD-H1; platelet receptor Gi24; PP2135; RIKEN cDNA 4632428N05 gene; SISP1; sisp-1; Stress-induced secreted protein-1; UNQ730/PRO1412; V-domain Ig suppressor of T cell activation; VISTA; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; VSIR; V-type immunoglobulin domain-containing suppressor of T-cell activation
Rat
100 μg
Primary
Mouse
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.