Learn More
VISTA Monoclonal Antibody (MH5A), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Affymetrix eBioscience 46-5919-82
Code : NEW
Additional Details : Weight : 0.09500kg
Description
VISTA is a homolog of PD-1 (PD-1H). PD-1H is a 309 aa type I transmembrane protein that belongs to the CD28 superfamily. This cell surface receptor is expressed on nearly all hematopoietic cells except B cells. VISTA negatively regulates T cell responses by suppressing early TCR activation, inhibiting cell proliferation, and suppressing TH1 cytokine production.
VISTA (V-domain Ig containing suppressor of T-cell activation), also known as B7-H5 or PD-1H, is a product of the VSIR gene. Although VISTA bears sequence homology to PD-L1 it has a different expression pattern. In both mouse and humans, VISTA is expressed predominantly on hematopoietic cells with the highest densities on myeloid cells, and lower levels on T cells. It is usually absent on B cells. VISTA expressed by antigen presenting cells has been shown to directly suppress proliferation and cytokine production of CD4 and CD8 T cells, making it a target for cancer immunotherapy. The reported ligands of VISTA include CD28H, VSIG-3 and VSIG-8.Specifications
VISTA | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q9D659 | |
VSIR | |
Affinity chromatography | |
RUO | |
74048 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MH5A | |
PerCP-eFluor 710 | |
VSIR | |
4632428N05Rik; B7H5; B7-H5; C10orf54; C1H10orf54; C28H10orf54; C4H10orf54; chromosome 10 open reading frame 54; DD1alpha; Death Domain1alpha; Dies1; differentiation of ESCs 1; GI24; PD-1H; PDCD1 homolog; PD-H1; platelet receptor Gi24; PP2135; RIKEN cDNA 4632428N05 gene; SISP1; sisp-1; Stress-induced secreted protein-1; UNQ730/PRO1412; V-domain Ig suppressor of T cell activation; VISTA; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; VSIR; V-type immunoglobulin domain-containing suppressor of T-cell activation | |
Armenian Hamster | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
For Research Use Only.