missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VISTA/B7-H5/PD-1H Rabbit anti-Mouse, Clone: 13F3, Novus Biologicals™
Rabbit Monoclonal Antibody
£200.00 - £463.00
Specifications
| Antigen | VISTA/B7-H5/PD-1H |
|---|---|
| Clone | 13F3 |
| Concentration | 1 mg/ml |
| Dilution | Flow Cytometry, ELISA, Block/Neutralize |
| Applications | Flow Cytometry, ELISA, Neutralization |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18371972
|
Novus Biologicals
NBP3-12054-0.025MG |
0.025 mg |
£200.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18343523
|
Bio-Techne
NBP3-12054-0.2MG |
0.2 mg |
£463.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VISTA/B7-H5/PD-1H Monoclonal antibody specifically detects VISTA/B7-H5/PD-1H in Mouse samples. It is validated for Flow Cytometry, ELISA, NeutralizationSpecifications
| VISTA/B7-H5/PD-1H | |
| 1 mg/ml | |
| Flow Cytometry, ELISA, Neutralization | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cardiovascular Biology, Immunology | |
| PBS | |
| 64115 | |
| IgG λ | |
| Protein A purified |
| 13F3 | |
| Flow Cytometry, ELISA, Block/Neutralize | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR | |
| This antibody was raised by immunising Armenian hamsters with EL4 cells overexpressing VISTA/B7-H5/PD-1H-RFP, and VISTA/B7-H5/PD-1H-Ig fusion protein. | |
| Primary | |
| Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title