missing translation for 'onlineSavingsMsg'
Learn More

VISTA/B7-H5/PD-1H Armenian Hamster anti-Mouse, Clone: MH5A, Novus Biologicals™

Product Code. 18374793 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
Unit Size:
0.20mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18374793 0.025 mg 25µg
18359891 0.2 mg 0.20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18374793 Supplier Novus Biologicals Supplier No. NBP3119690.025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Armenian Hamster Monoclonal Antibody

VISTA/B7-H5/PD-1H Monoclonal antibody specifically detects VISTA/B7-H5/PD-1H in Mouse samples. It is validated for ELISA, Immunohistochemistry, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen VISTA/B7-H5/PD-1H
Applications Neutralization
Classification Monoclonal
Clone MH5A
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry, Block/Neutralize
Formulation PBS
Gene Alias B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR
Host Species Armenian Hamster
Immunogen The antibody was generated by immunising Armenian hamsters with mVISTA/B7-H5/PD-1HIg.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 64115
Target Species Mouse
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Isotype IgG λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.